Thermoelectric Generator Demonstrator
Catalog #: 470163-908
Supplier: American Scientific
Specifications
- Description:Thermoelectric Generator Demonstrator
- Cat. no.:470163-908
- Supplier No.:1000-12
Specifications
About this item
Bring the 2nd Law of Thermodynamics to life.
- Two cups included
- Comes with a Teacher's Guide
This device can act as a refrigerator or a heat engine. Immerse the two aluminum legs in baths of different temperatures, and produce electrical energy that turns the turbine! Unplug the banana jacks and measure the voltage output with a multimeter.
Customers Who Bought This Also Bought