Air Puck Hovercraft
Catalog #: 470163-906
Supplier: American Scientific
Specifications
- Description:Air puck hovercraft
- Height:2¼"
- Cat. no.:470163-906
- Supplier No.:3569-00
- Diameter:7½"
Specifications
About this item
Demonstrate Newton’s Laws and the conservation of energy and momentum with the amazing new Air Puck!
- Teacher's guide included, with NGSS alignment
- Requires 4 × AA batteries, not included
The Air Puck hovercraft is perfectly balanced and glides on a cushion of air for nearly frictionless motion across any smooth surface. Sturdy bumpers will survive collisions with walls, shoes, or other Air Pucks. Use it to demonstrate Newton's First Law, uniform linear motion. Or, collide two Air Pucks to demonstrate conservation of momentum in two dimensions!
Ordering information: Mini hovercraft launcher is recommended but not included.
Customers Who Bought This Also Bought