Radiation Cans
Catalog #: 470163-902
Supplier: American Scientific
Specifications
- Description:Radiation Cans, Set of 3
- Cat. no.:470163-902
- Supplier No.:AR-725
Specifications
About this item
Radiation cans for demonstrating solar energy and heat transfer.
- The cans are finished with silver, black, and white exteriors
- Set of three 500ml cans
Just add water and a thermometer to each of the three cans, place in the sun or near a bright light bulb and watch the temperatures rise.
Customers Who Bought This Also Bought