Diaphragm with 4 Double Slits of Different Spacings
Catalog #: 470163-762
Supplier: American 3B Scientific
Specifications
- Description:Diaphragm with 4 Double Slits of Different Spacings
- Cat. no.:470163-762
- Supplier No.:U14101
Specifications
About this item
Photographic diffraction objects in a slide frame.
- Dimensions: 50x50 mm²
- Number of slits: 4 sets of 2
- Slit width [mm]: 0.15
- Slit spacing [mm]: 0.25; 0.50; 0.75; 1.00
Customers Who Bought This Also Bought