Activity Series Kit
Catalog #: 470163-070
Supplier: Ward's Science
Specifications
- Description:Activity Series Kit
- No. of Lab Groups:15
- Cat. no.:470163-070
- Supplier No.:IS8002
Specifications
About this item
Oxidation and Reduction of Metals
- Classic AP Chemistry Experiment
- Determine Which Metal is the Most Reactive
- Instructions Included
An activity series of metals is a table of metals arranged in the order of their decreasing chemical activity or the ease at which the metal will give up one or more electrons to form positive ions. This table is similar to the electrochemical series of elements. For example if you take the group of metals magnesium, mercury and nickel, magnesium is the most reactive and mercury the least. To empirically determine which of these metals is more reactive, place a piece of the metal in a salt solution of the other. The more reactive metal will replace the less reactive metal and the less reactive will appear in the solid form. The reactive metal has been oxidized; the less reactive metal has been reduced.
Ordering information: Includes Copper Metal, Zinc Metal, Magnesium Metal, Lead Metal, Copper Nitrate Solution, Zinc Nitrate Solution, Magnesium Nitrate Solution, Lead Nitrate Solution, Potassium Bromide, Potassium Bromate, Sulfuric Acid Solution, Chlorine Water, Iodine Water, Sodium Bromide Solution, Potassium Iodide Solution, Sodium Chloride Solution, Mineral Oil, 1 Empty Bottle (for Bromine Water), Teacher's Guide and Student Copymasters.
Caution: Classic AP Chemistry Experiment
Determine Which Metal is the Most Reactive
Instructions Included
Customers Who Bought This Also Bought