CENCO® Assorted Ball Set
Catalog #: 470164-716
Supplier: United Scientific Supplies
Specifications
- Description:CENCO® Assorted Ball Set
- Supplier No.:DBLST12-A
Specifications
About this item
Comprehensive and High Quality.
- Packed in plastic storage box
- Set of 12
A set with 12 balls in all, includes 25mm diameter drilled balls of brass, aluminum, wood, steel, copper and lead; two 25mm solid steel balls, one plain and one with bifilar suspension lugs; 38 mm balls of brass, steel and lead with hooks; and one 25mm solid cork ball.
Customers Who Bought This Also Bought