Order Entry
ContactUsLinkComponent
Anti-MMP13 Rabbit Polyclonal Antibody
Anti-MMP13 Rabbit Polyclonal Antibody
Catalog #: 80089-038
Supplier:  Abgent
MMP13 C-TERM ANTIBODY 0.1MG
Anti-MMP13 Rabbit Polyclonal Antibody
Catalog #: 80089-038
Supplier:  Abgent
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 13
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Antigen symbol:
    MMP13
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    400 µL
  • Western blot:
    Yes
  • Epitope:
    C-Terminal (335–349 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    MMP-13|CLG3|Collagenase 3|matrix metallopeptidase 13|MANDP1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Cat. no.:
    80089-038
  • Supplier No.:
    AP6197A

Specifications

About this item

Type: Primary
Antigen: MMP13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (335–349 aa)
Host: Rabbit
Isotype:
Reactivity: Human

Customers Who Bought This Also Bought