About this item
Avi Tag Is A Biotin-Acceptor Peptide, Glndifeaqkiewhe. The 15-Residue Peptide Served As A Substrate Mimic For Biotin Ligase (Bira), Which Usually Recognizes The Much Larger Protein Domain. Avi Tag Is A Useful Tool In Analysis Of Avi Fusion Proteins.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 89493-824
- Antigen name:
- Avi-Tag
- Clonality:
- Monoclonal
- Host:
- Mouse