To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:IgG
- Clonality:Monoclonal
- Clone:4H3G6
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:IgG
- Conjugation:Unconjugated
- Size:1 mg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage temperature:The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 1 mg/ml
- Shipping temperature:Ice
- Purification:Protein A affinity column
- Cat. no.:89494-612
- Packaging:Vial
- Supplier No.:A01730
Specifications
About this item
Mouse IgG2b control (Whole Molecule), Purified is suitable to be used as control, standard, blocking agent, or coating protein in a variety of assays, including ELISA, immunobloting (Dot blot and Western blot), immunoprecipitation, immunodiffusion, and immunoelectrophoresis. It may also be used as antigen or ligand in immunochemical conjugation reaction. GenScript Mouse IgG2b control (Whole Molecule)is highly purified from normal mouse ascites through Protein A affinity column and produced from the hybridoma resulting from fusion of SP2/0-Ag14 myeloma and B-lymphocytes.
- High specificit
- High affinity
- High stability
GenScript offers a wide selection of polyclonal and monoclonal loading control antibodies for α-tubulin and β-actin for several species. HRP- and biotin-conjugated versions of these antibodies are also available. GenScript also offers a series of isotype control antibodies that can be used in your research.
Type: Control
Antigen: IgG2 beta
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: