To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:IgG
- Clonality:Polyclonal
- Host:Mouse
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:IgG
- Conjugation:Unconjugated
- Size:1 mg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage temperature:The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 1 mg/ml
- Shipping temperature:Ice
- Purification:Protein A affinity column
- Cat. no.:89494-608
- Packaging:Vial
- Supplier No.:A01007
Specifications
About this item
GenScript Purified Mouse IgG (Whole Molecule) Control is highly purified from normal mouse serum through Protein A chromatography.GenScript Purified Mouse IgG (Whole Molecule) Control is suitable to be used as control, standard, blocking agent, or coating protein in a variety of assays, including ELISA, immunobloting (Dot blot and Western blot), immunoprecipitation, immunodiffusion, and immunoelectrophoresis.It may also be used as antigen or ligand in immunochemical conjugation reaction.
- High specificit
- High affinity
- High stability
GenScript offers a wide selection of polyclonal and monoclonal loading control antibodies for α-tubulin and β-actin for several species. HRP- and biotin-conjugated versions of these antibodies are also available. GenScript also offers a series of isotype control antibodies that can be used in your research.
Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: