To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Tag
- Antigen name:GST Tag
- Clonality:Monoclonal
- Clone:2F10B9
- Host:Mouse
- Isotype:IgG1
- Antigen symbol:GST Tag
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:GSTP1-1|PI|DFN7|GST3|GSTP1|FAEES3
- Storage buffer:PBS, pH 7.4, containing 1% BSA and 0.01% thimerosal
- Storage temperature:The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant GST protein (26 kD)
- Purification:Protein A affinity column
- Cat. no.:89493-656
- Packaging:Vial
- Supplier No.:A00866-100
Specifications
About this item
THE™ GST antibody [HRP], mAb, Mouse recognizes C-terminal, N-terminal and internal tagged GST fusion proteins.
- Comprehensive pool
- Against 24 tags
- Available in gram level bulk quantities
- Various conjugates for hot tags detection
GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.
Type: Tag
Antigen: GST
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: