About this item
ASEPTI-CLEANSE® is a hands-free dispensing system with the most advanced infrared sensor which can be set to dispense approximately 1, 3 or 5 mL of solution
- Dispenses a pre-measured dose to the hand without user contact
- Photo-eye operated
- Water resistant design
- Operates on 4 D-Cell Batteries or 110 V direct connections
Asepti-Cleanse® has been designed by VAI as the most advanced Infrared Sensor Dispensing System available in the pharmaceutical and biotech industry. The dispensing unit is designed to
meet the requirements of cGMP clean room operations.
Excellent choice for gowning rooms and aseptic manufacturing areas.