To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Format:Spin column
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:Zyppy™ Plasmid Miniprep Kit
- Size:100 preps
- Description:Zyppy™ Plasmid Miniprep Kit
Specifications
About this item
The Zyppy™ Plasmid Purification Kits feature a pellet-free procedure for the fastest purification of plasmid DNA.
- The fastest, easiest miniprep available for purifying high-quality plasmid DNA
- Pellet-free procedure omits conventional cell pelleting and re-suspension steps
- DNA quality is sufficient for cloning and sequencing
The Zyppy™ Plasmid Kits feature a pellet-free modified alkaline lysis method that bypasses bacterial culture centrifugation and re-suspension steps common to classical plasmid preparation procedures. Simply add the uniquely formulated 7X Lysis Buffer directly to bacterial culture, neutralize, and then purify using the provided Zymo-Spin column technology. Additionally, the innovative coloured buffers included in the kit permit error-free visualization and identification of complete bacterial cell lysis and neutralization. The Zyppy™ Plasmid Kits are the fastest and easiest method available to separate plasmid DNA from E. Coli efficiently. The plasmid DNA is of the highest quality, and is well suited for use in bacterial transformation, restriction endonuclease digestion, DNA ligation, PCR, transcription, sequencing, and other sensitive downstream applications.