To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Bromodeoxyuridine
- Clonality:Monoclonal
- Clone:2B1
- Host:Mouse
- Isotype:IgG1
- Reactivity:Multiple
- Antigen symbol:BrdU
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Form:Liquid
- Storage buffer:PBS, pH 7.4, containing 10mg/ml sucrose and 1.0mg/ml sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:Shipped on Blue Ice
- Immunogen:Iododeoxyuridine-Ovalbumin.
- Purification:purified by Protein A affinity chromatography
- Cat. no.:10049-410
- Supplier No.:ADI-MSA-200-E
Specifications
About this item
The thymidine analogue BrdU (5-Bromo-2-Deoxyuridine), a derivative of uridine, can substitute for thymidine during DNA synthesis. The detection of BrdU incorporation into DNA provides a common method to quantify newly synthesized DNA and identify cells in the S-phase of the cell cycle. The use of BrdU incorporation in proliferation assays facilitates the study of DNA repair, sister chromatid exchange, and the cytokinetics of normal and neoplastic cells.
Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: 2B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: pan