Weighing Dishes, Wheaton
Catalog #: 30617-222
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Dimensions:Dia.×H 20×8 mm (⁴⁄₅׳⁄₁₀")
- Material:Aluminium
- Supplier No.:370792
Specifications
About this item
This weighing dishes are made from aluminum which is ideal for milligram weighing.
- For use with liquids or solids
- Smooth surface ensures complete transfer
- Use of balance-type forceps enables easy grasping
Customers Who Bought This Also Bought