Order Entry
ContactUsLinkComponent
Anti-CD44 Rabbit Polyclonal Antibody
Anti-CD44 Rabbit Polyclonal Antibody
Catalog #: 10209-728
CD44 antibody
Anti-CD44 Rabbit Polyclonal Antibody
Catalog #: 10209-728
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cluster of Differentiation 44 antigen
  • Clonality:
    Polyclonal
  • Isotype:
    IgG
  • Antigen symbol:
    CD44
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    MDU2|HUTCH-I|LHR|MC56|HCAM|Pgp1|CDW44|MIC4|ECMR-III|HCELL|MDU3|IN|CSPG8
  • Amino acid number:
    728 to 742
  • Sequence:
    DETRNLQNVDMKIGV
  • Storage temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human CD44 (728-742aa DETRNLQNVDMKIGV), different from the related mouse and rat sequences by one amino acid.
  • Purification:
    Purified by immunogen affinity purification
  • Cat. no.:
    10209-728
  • Supplier No.:
    PA1021-2

Specifications

About this item

Rabbit IgG polyclonal antibody for CD44 antigen(CD44) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CD44
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity:

Customers Who Bought This Also Bought