To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Caldesmon (smooth)
- Clonality:Monoclonal
- Clone:CD-7
- Gene ID:Human: 800, Mouse: 109624, Rat: 25687
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rabbit,Human,Mouse
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Liquid
- UniProtKB:Human: Q05682, Mouse: Q8VCQ8, Rat: Q62736
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Human uterus smooth muscle extract.
- Cat. no.:10209-324
- Supplier No.:MA1009
Specifications
About this item
Mouse IgG monoclonal antibody for Caldesmon (smooth), caldesmon 1 (CALD1) detection. Tested with WB, IHC-F in Human;mouse;rat. No cross reactivity with other proteins.
Caldesmon is a potential actomyosin regulatory protein found in smooth muscle and nonmuscle cells. The predicted smooth-muscle polypeptide is 793 amino acids long. The high molecular weight caldesmon(h-CaD) is predominantly expressed in smooth muscles, whereas the low molecular weight caldesmon(l-CaD) is widely distributed in nonmuscle tissues and cells. Hayashi et al.(1992) demonstrated that the human CDM gene is composed of 14 exons.
Mouse ascites fluid, 1.2% sodium acetate, 2mg BSA, with 0.01mg NaN3 as preservative.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: Caldesmon (smooth)
Clonality: monoclonal
Clone: CD-7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rabbit