To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:NAPDH oxidase 2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NOX2
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Cytochrome b(558) subunit beta|Neutrophil cytochrome b 91 kDa polypeptide|NADPH oxidase 2|cytochrome b-245|chronic granulomatous disease|Superoxide-generating NADPH oxidase heavy chain subunit|CGD|Heme-binding membrane glycoprotein gp91phox|GP91-PHOX|CYBB|GP91 PHOX|GP91-1
- Amino acid number:442 to 459
- Sequence:YWLCRDTHAFEWFADLLQ
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human NOX2 (442-459aa YWLCRDTHAFEWFADLLQ), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-202
- Supplier No.:PA1667
Specifications
About this item
Rabbit IgG polyclonal antibody for Cytochrome b-245 heavy chain(CYBB) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: NOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat