To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:MAP Kinase, Activated
- Clonality:Monoclonal
- Clone:IL-13
- Host:Mouse
- Isotype:IgG1
- Reactivity:Yeast,Human,Rat,Mouse
- Antigen symbol:MAP Kinase, Activated
- Conjugation:KLH (Keyhole Limpet Hemocyanin)
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Modification:phospho-Glu, phospho-Val
- Sequence:HTGFLTpEYpVAT
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide containing 11 amino acids, HTGFLTpEYpVAT, corresponding to the phosphorylated form of ERK-activation loop conjugated to KLH.
- Cat. no.:10207-930
- Supplier No.:MA1055
Specifications
About this item
Mouse IgG monoclonal antibody for MAP Kinase, Activated (Diphosphorylated ERK-1&2), mitogen-activated protein kinase 1; mitogen-activated protein kinase 3 (MAPK1; MAPK3 ) detection. Tested with WB, IHC-P, ICC in Human;mouse;rat;yeast. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: MAP Kinase, Activated
Clonality: monoclonal
Clone: IL-13
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Yeast