To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cluster of differentiation 8
- Clonality:Monoclonal
- Clone:CA-8
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Antigen symbol:CD8
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:CD8alpha|p32|Leu-2|CD8a|MAL|CD8|CD8b|CD8beta
- Storage temperature:–20 °C
- Immunogen:Human thymocytes followed by peripheral blood T cells.
- Cat. no.:10206-192
- Supplier No.:MA1017
Specifications
About this item
Mouse IgG monoclonal antibody for CD8, CD8a molecule; CD8b molecule (CD8A; CD8B) detection. Tested with IHC-F in Human. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: CA-8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: