To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Actin Alpha 1, Skeletal Muscle
- Clonality:Monoclonal
- Clone:AC-40
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Chicken,Human,Rat,Mouse
- Antigen symbol:ACTA1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:ACT1|alpha skeletal muscle Actin|ASMA|alpha Actin 1|NEM2|ACTA|NEM1
- Storage temperature:–20 °C
- Immunogen:Synthetic actin C-terminal peptideSer-Gly-Pro-Ser-Ile-Val-His-Arg-Lys-Cys-Phe,attached to a Multiple Antigen Peptide (MAP) backbone.
- Cat. no.:10206-118
- Supplier No.:MA1000
Specifications
About this item
Mouse IgG monoclonal antibody for Actin detection. Tested with WB, IHC-F, ICC in Human;mouse;rat;chicken. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: ACT1
Clonality: Monoclonal
Clone: AC-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Chicken