To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:gamma-Tubulin
- Clonality:Monoclonal
- Clone:IMD-20
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Antigen symbol:TUBG
- Conjugation:KLH (Keyhole Limpet Hemocyanin)
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Amino acid number:38 to 53
- Storage temperature:–20 °C
- Immunogen:Synthetic gamma-tubulin peptide (amino acids 38-53), conjugated to KLH.
- Cat. no.:10205-934
- Supplier No.:MA1114
Specifications
About this item
Mouse IgG monoclonal antibody for gamma-Tubulin, golgi-associated, gamma adaptin ear containing, ARF binding protein 1; golgi-associated, gamma adaptin ear containing, ARF binding protein 2; golgi-associated, gamma adaptin ear containing, ARF binding protein 3 (GGA1; GGA2; GGA3 ) detection. Tested with WB in Human,rat. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: gamma-Tubulin
Clonality: monoclonal
Clone: IMD-20
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat