To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Nitric Oxide Synthase 1
- Clonality:Monoclonal
- Clone:N1
- Host:Mouse
- Isotype:IgG1
- Reactivity:Pig,Goat,Human,Rat
- Antigen symbol:NOS1
- Conjugation:Unconjugated
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:N-NOS|Peptidyl-cysteine S-nitrosylase NOS1|Constitutive NOS|NC-NOS|Nitric oxide synthase|bNOS|brain|NOS1|NOS type I|nNOS|Neuronal NOS
- Amino acid number:1 to 181
- Storage temperature:–20 °C
- Immunogen:Recombinant neuronal NOS fragment (amino acids 1-181) from rat brain.
- Cat. no.:10205-896
- Supplier No.:MA1072
Specifications
About this item
Mouse IgG monoclonal antibody for Nitric Oxide Synthase, Brain (1-181) NOS1, nitric oxide synthase 1 (neuronal) (NOS1) detection. Tested with WB in Human;rat;goat;pig. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: NOS1
Clonality: Monoclonal
Clone: N1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Goat, Pig