BetaBasic™ 18 3 and 5 µm HPLC Columns, Thermo Scientific
BetaBasic columns are highly efficient and reproducible, and feature a 150 Å pore size for small molecules, peptides and protein digests.
Superb pH stability.
- Catalog No:
- 10046-434
- 10046-392
- 10046-376
- Description:
- 10046-434
- For Uniguard Direct-connection and Stand Alone Holder
- 10046-376
- Carbon load:
- 13%
- 13%
- 13%
- Endcapped:
- Yes
- Yes
- Yes
- pH range:
- 2 - 12
- 2 - 12
- 2 - 12
- Surface area:
- 200 m²/g
- 200 m²/g
- 200 m²/g
- USP code:
- L1
- L1
- L1
Frequently Bought Together