Specifications
- Volume:10 µl
- Needle Length:57 mm (21/4")
- Gauge:26s
- Supplier No.:365D0291
Specifications
About this item
Achieve accurate, precise, and reproducible injections easily using Thermo Scientific™ GC Syringes for Thermo Scientific™ TriPlus™ RSH Autosampler
The robust, field-proven designs are optimized for septum piercing. They are made of borosilicate glass with metal flanges of either stainless steel or nickel-plated brass and cover a range of volumes.