Sample Loops for Rheodyne® Sample Injectors, IDEX Health & Science
Supplier: Rheodyne
Certificates
Certificates
About this item
Customers Who Bought This Also Bought
Silver nitrate 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$297.60 to $1,622.45
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$64.40 to $734.93
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
$386.44 to $1,545.78
Trisodium phosphate dodecahydrate 98.0-102.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
$147.08 to $1,469.62
Acetic acid glacial 99.7-100.5% ACS, meets analytical specification of USP, FCC, VWR Chemicals BDH®
Supplier: BDH
$720.66 to $3,438.57
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15 × 100 mm plate, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
$42.99