Manual 2-Position Switching Valves, Rheodyne®, IDEX Health & Science
Catalog #: 97058-314
Supplier: Rheodyne
Specifications
- Description:2-position, 6-port switching valve, large bore, 10-32 ports, 1.0 mm stator passage diameter, Stainless steel, For 1/16" OD tubing
- Cat. no.:97058-314
- Supplier No.:7000L
Specifications
About this item
Applicable for sample injection, column switching and sample enrichment applications.
Customers Who Bought This Also Bought