Storage Vials and Closures
Supplier: Thermo Scientific
Certificates
About this item
These storage screw thread vials are manufactured from superior quality 33 expansion borosilicate clear Type 1, Class A or 51A amber Type 1, Class B glass.
- PTFE-lined solid-top storage caps
- Not autoclavable
- PTFE on film/foam packing offers broad chemical resistance
The storage vials range from 2 to 40 ml.