Memphis UltraTech® Gloves, Air Infused, MCR Safety
Supplier: MCR Safety
Certificates
About this item
UltraTech® provides ultra comfort, ultra fit, and ultra protection.
- The seamless nylon shell offers outstanding breathablility and comfort
- The polyurethane palm and finger coating provides grip,puncture resistance, abrasion resistance and dexterity
Great for small parts handling. The 9694 features a 15 gauge white nylon shell and gray air infused nitrile coating on the palm and fingertips.
Ordering information: These gloves are polybagged by the pair.
Specifications
- Catalog No:
- 80081-728
- 80081-732
- 80081-730
- 80081-736
- 80081-734
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Finish:
- Coated Palm
- Coated Palm
- Coated Palm
- Coated Palm
- Coated Palm
- Gauge:
- 15 Gauge
- 15 Gauge
- 15 Gauge
- 15 Gauge
- 15 Gauge
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist