FlexTherm® Gloves, MCR Safety
Supplier: MCR Safety
Certificates
About this item
Flex Therm options give all of the benefits of flex tuff series of gloves, but with the addition of Thermal protection! The 9690 is made of a an acrylic/polyester/cotton string knit shell with a gray latex palm coating.
- Extra thermal protection
- High comfort level
- Allows manual dexterity
- For heavier duty requirements
These gloves can provides a grip on staying warm.
Ordering information: These gloves are polybagged by the pair.
Specifications
- Catalog No:
- 80081-718
- 80081-716
- 80081-714
- 80081-712
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Finish:
- Coated Palm
- Coated Palm
- Coated Palm
- Coated Palm
- Gauge:
- 10 Gauge
- 10 Gauge
- 10 Gauge
- 10 Gauge
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist