Regular Weight String Knit Gloves, MCR Safety
Supplier: MCR Safety
Certificates
About this item
Our multi-purpose string knit gloves come in various blends and weights.
- Color coded hem to identify sizes
- Made with FDA accepted materials
- Overedge hem
- Reversible pattern
Economy, regular, and heavy weight gauge offerings are available. These gloves are suitable for general hand protection in light duty applications. The 9500 series features a regular weight 7 gauge white cotton/polyester blend and overedge hem that is excellent for laundering.
Specifications
- Catalog No:
- 89135-722
- 89135-726
- 89135-724
- 89135-730
- 80081-646
- 80081-644
- 80081-648
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Gauge:
- 7 Gauge
- 7 Gauge
- 7 Gauge
- 7 Gauge
- 7 Gauge
- 7 Gauge
- 7 Gauge
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist