Cotton Inspectors' Gloves, MCR Safety
Supplier: MCR Safety
Certificates
About this item
Smooth, thin and cool nylon inspection gloves good for low lint applications.
- Two piece reversible
- Breathable fabric helps keep hands cool
- Comfortable and light weight
- Straight thumb
- Thin white nylon gloves
They can be worn alone or used as glove liners. The 8700 glove is made of 100% white nylon.
Specifications
- Catalog No:
- 80081-610
- 80081-608
- 80081-614
- 80081-612
- 80081-606
- 80081-604
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Cuff style:
- Slip-on
- Slip-on
- Slip-on
- Slip-on
- Slip-on
- Slip-on