Cotton Gloves, Brown Jersey, MCR Safety
Supplier: MCR Safety
Certificates
About this item
Brown jersey gloves have been used for generations as a light weight, comfortable option for light duty.
- The 7100 is a clute pattern glove
- The cotton/polyester blend is excellent for laundering
Brown jersey gloves are available in many weights, patterns, and even some with dots for grip.
Specifications
- Catalog No:
- 80081-596
- Ambidextrous/Hand-specific:
- Ambidextrous
- Cuff style:
- Knit wrist