Memphis NeoMax™, Double Dipped Neoprene Gloves, Industrial Grade, MCR Safety
Supplier: MCR Safety
Certificates
About this item
Not only does the NeoMax have a great name, but it offer outstanding protection.
- Made of green double dipped neoprene
- 100% cotton brushed interlock liner
- Component materials comply with all federal regulations for food contact
The sandy finish helps get a grip and the brused interlock lining is treated with ActiFresh® that will prevent bacterial build up and resist molds and mildew. Go to the Max with NeoMax™.
Specifications
- Catalog No:
- 80081-584
- 80081-578
- 80081-576
- 80081-582
- 80081-580
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet