Chem-Tech™ Gloves, Neoprene Over Latex, Industrial Grade, MCR Safety
Supplier: MCR Safety
Certificates
About this item
Our neoprene gloves are thick, waterproof, and an effective replacement for latex and vinyl products.
- Chem-Tech 5400S series features
- 28 mil blue neoprene over yellow latex
- Flock lining and straight cuff
Neoprene is a synthetic rubber that maintains flexibility over a wide range of temperatures.
Ordering information: These gloves are polybagged by the pair.
Specifications
- Catalog No:
- 80081-486
- 80081-488
- 80081-492
- 80081-494
- 80081-496
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Liner material:
- Flock
- Flock
- Flock
- Flock
- Flock
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- Straight
- FDA regulated:
- +
- +
- +
- +
- +