Specifications
- Sensitivity:30 ng/mL (range 3-810 ng/mL)
- Cat. no.:CA10551-284
- Supplier No.:KIT127-0001
- No. of Tests:96 Wells
Specifications
About this item
Sensitive (30 ng/ml) ELISA for the quantitative determination of host cell protein contamination in bulk products expressed in E
For the quantitative determination of contamination with E.coli host cell protein.
The E. coli host cell protein ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of E. coli host cell protein contamination with results in 3 hours
coli expression systems.