M006 Nylon Glove Liner, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
This reusable half-finger liner contains 100% continuous nylon filament.
- Ambidextrous
- Reusable
- For critical environments
- Washing machine friendly up to 30 times without shrinking
It can be washed 30 times without shrinking, reducing cost of replacement and waste disposal.
Ordering information: These gloves are sold by the dozen pairs.
Specifications
- Catalog No:
- 32935-566
- 32935-568
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous