To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Assay Type:Competitive
- Sensitivity:4.31pg/mL (range 9.8 - 10,000pg/mL)
- Target protein:11-dehydro-TXB2
- Cat. no.:89141-140
- Supplier No.:900-092
- No. of Tests:96 Wells
Specifications
About this item
For the quantitative determination of 11-dehydro-TXB2 in culture supernatants and urine from any species
The 11-dehydro-TXB2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405nm. This kit offers ultrasensitive quantification of 11-d-TXB2 in a variety of sample matrices with negligible reactivity with related molecules in a convenient 96-well format offers higher throughput analysis compared to conventional analytical methods.
Cited samples include serum.