About this item
Ideal for the determination for pH, mV, Ion concentration, and Dissolved Oxygen based on optical RDO® (Rugged Dissolved Oxygen) technology.
- Multiparameter
- Store up to 500 data points
- Five-point calibration
- Intelligent sensor management system (ISM®)
These meters are essential for special analyses, particularly in the in the environmental, chemical and food & beverage industry. The meter stores up to 500GLP measurements, with Date/Time, Sensor ID and Serial number, user and Sample ID. The meter features five point pH calibration with a choice of seven predefined and one user-defined buffer group. The revolutionary Intelligent Sensor Management system (ISM®) offers the user an even greater level of security. The calibration data is stored on the chip in the sensor and is used automatically when the sensor is connected to the meter. This ensures that the meter always uses the latest calibration data.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 89186-072
- 89186-066
- 89186-068
- 89186-070
- ISE accuracy:
- ±0.5%
- ±0.5%
- ±0.5%
- ±0.5%
- ISE concentration range:
- 1.00E⁻⁹ - 9.99E⁺⁹
- 1.00E⁻⁹ - 9.99E⁺⁹
- 1.00E⁻⁹ - 9.99E⁺⁹
- 1.00E⁻⁹ - 9.99E⁺⁹
- DO range:
- 0.00 - 50.0 ppm (mg/L)
- 0.00 - 50.0 ppm (mg/L)
- 0.00 - 50.0 ppm (mg/L)
- 0.00 - 50.0 ppm (mg/L)
- Temperature resolution:
- 0.1 °C
- 0.1 °C
- 0.1 °C
- 0.1 °C
- Pressure resolution:
- 1 mbar
- 1 mbar
- 1 mbar
- 1 mbar
- Pressure accuracy:
- ±1
- ±1
- ±1
- ±1
- Operating temperature range:
- 0…40 °C (32…104 °F)
- 0…40 °C (32…104 °F)
- 0…40 °C (32…104 °F)
- 0…40 °C (32…104 °F)