To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Protein Tyrosine Phosphatase Receptor Type C
- Clonality:Monoclonal
- Clone:RA3-6B2
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse
- Antigen symbol:PTPRC
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:500 µg
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Storage buffer:0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.09% (w/v) Sodium Azide, 0.1% Gelatin
- Storage temperature:+4 °C
- Concentration:0.5mg/ml by UV absorbance at 280 nm
- Shipping temperature:Wet Ice
- Immunogen:Anti-CD45R (B220) Antibody (Monoclonal) was produced by repeated immunizations with CD45R antigen.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Cat. no.:CA200-502-N84
- Supplier No.:200-502-N84
Specifications
About this item
Anti-CD45R (B220) is useful for Flow Cytometry, Immunoprecipitation, immunochemistry, Immunofluorescence, and Western Blot using mouse spleen cells, or an appropriate cell type (where indicated).
Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze!
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Mouse