To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Alkaline Phosphatase, Intestinal
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:ALPI
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Size:25 µL
- Western blot:Yes
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Gentamicin Sulfate. Do NOT add Sodium Azide!
- Concentration:1 mg/ml by UV absorbance at 280 nm
- Tested applications:Fluoresence ELISA
- Cat. no.:CA10037-020
- Supplier No.:200-403-142S
Specifications
About this item
Anti-Alkaline Phosphatase has been assayed against 1.0 µg of Alkaline Phosphatase (Human Intestine) by immunoblot using TMB 3,3',5,5'Tetramethylbenzidine as a substrate at room temperature.
Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000;
Do not add Sodium Azide!
Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human