To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Carbonic Anhydrase 2
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:CA2
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Size:25 µL
- Western blot:Yes
- Antigen synonyms:CAC|Carbonic dehydratase|CA-II|Carbonate dehydratase II|HEL-76|Carbonic anhydrase II|Carbonic anhydrase C|Car2|Carbonic anhydrase B|CAII
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Gentamicin Sulfate. Do NOT add Sodium Azide!
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Tested applications:Fluoresence ELISA
- Purification:multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above
- Cat. no.:CA10037-004
- Supplier No.:200-403-136S
Specifications
About this item
Anti-Carbonic anhydrase II has been assayed against 1.0 µg of Carbonic Anhydrase II (Human Erythrocytes) in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])
Recommended Dilutions: ELISA: 1:500 - 1:2500; Western Blot: 1:250 - 1:1000; Immunochemistry: 1:100 - 1:500;
Do not add Sodium Azide!
Type: Primary
Antigen: CA2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human