To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Tag
- Antigen name:Galactosidase Beta 1
- Clonality:Polyclonal
- Host:Rabbit
- Antigen symbol:GLB1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Size:25 µL
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:PPCA|PPGB|RP3-337O18.1|NGBE|GLB2|Beta-Gal|GSL
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide
- Concentration:1.0mg/ml by UV absorbance at 280 nm
- Tested applications:Fluoresence ELISA
- Purification:multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above
- Cat. no.:CA200-402-036S
- Supplier No.:200-402-036S
Specifications
About this item
Suitable for fluorescent western blotting and immunofluorescence.
Recommended Dilutions:
Type: Tag
Antigen: b-GAL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: