To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Calcitonin-related Polypeptide Alpha
- Clonality:Monoclonal
- Clone:18C4.B4.C2
- Host:Mouse
- Isotype:IgG1 lambda
- Reactivity:Human
- Antigen symbol:CALCA
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:CT|Calcitonin-related Polypeptide alpha|CGRP|CALC1|KC|CGRP1|CGRP-I|Calcitonin
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:Procalcitonin Antibody was produced in mice prepared by repeated immunizations with full-length recombinant human Procalcitonin Protein.
- Tested applications:Fluoresence ELISA
- Purification:Protein A purified
- Cat. no.:CA200-301-I40
- Supplier No.:200-301-I40
Specifications
About this item
Anti-Procalcitonin antibody is suitbale for use in ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:1000-1:20000; Western Blot: 1µg/ml;
Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: 18C4.B4.C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human