To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Clonality:Monoclonal
- Clone:eB144
- Host:Rat
- Reactivity:Mouse
- Antigen symbol:IgG
- Conjugation:DyLight® 680
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µL
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Polyethylene Glycol (PEG-8000)
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Cat. no.:CA18-4417-32
- Supplier No.:18-4417-32
Specifications
About this item
Fluorescent Mouse TrueBlot® Antibody DyLight® 680 is provided as a lyophilized powder. In order to conserve reagent, we recommend incubating the blots from minigels in sealed bags (removing as much air as possible) with minimal volume (2-3 mls).
Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: eB144
Conjugation: DyLight® 680
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse