About this item
Ideal for routine pH, mV, Ion concentration, and Dissolved oxygen measurements
- Multiparameter
- Store up to 500 data points
- Automatic endpoint and buffer recognition
- Five-point calibration
- Intelligent sensor management system (ISM®)
These units store up to 500 GLP data points with Time/Date, Sensor ID, User name, and Sample ID. Units feature automatic endpoint and buffer recognition, and five-point pH calibration with seven predefined and one user-defined buffer set. It is equipped with sophisticated new concept, Intelligent Sensor Management (ISM®). The display shows measured values and settings while continuously indicating electrode and battery status. Units are equipped with rotatable electrode clip that can be mounted on the left or the right. It allows single-handed measurement and saves space when the electrode is not being used.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA97025-858
- CA97025-860
- CA97025-856
- ISE accuracy:
- ±0.5%
- ±0.5%
- ±0.5%
- ISE concentration range:
- 1.00E-9 to 9.99E+9
- 1.00E-9 to 9.99E+9
- 1.00E-9 to 9.99E+9
- Temperature resolution:
- 0.1°C
- 0.1°C
- 0.1°C
- Temperature accuracy:
- ±0.2°C
- ±0.2°C
- ±0.2°C
- Pressure resolution:
- 1 mbar
- 1 mbar
- 1 mbar
- Pressure accuracy:
- ±1
- ±1
- ±1
- Operating temperature range:
- 0…40 °C (32…104 °F)
- 0…40 °C (32…104 °F)
- 0…40 °C (32…104 °F)