Shipper, EAS Combo, Therapak®
Catalog #: 10029-422
Supplier: Therapak, LLC
Specifications
- Description:EAS combination shipping system
- Cat. no.:10029-422
- Supplier No.:56886
Specifications
About this item
EAS Combination specimen shipping system features Therapak®'s standard small cooler (for frozen specimen section) nested underneath an insulated envelope (for ambient specimen section).
Frozen specimens are packed inside a 6x9" zip-style biohazard bag, including an Aqui-Pak™ segmented absorbent pouch that holds up to four tubes, then placed in the small insulated cooler (ID: 8 x 6 x 8”) with the appropriate amount of dry ice within a transport box (OD: 11 x 9 x 11”).
Ambient section includes 6x9" biohazard bag and Aqui-Pak™ segmented absorbent pouch plus an ambient gel wrap to maintain the temperature environment.
System includes: cushioning material, Exempt Animal Specimen label and biohazard label.
Add dry ice and sample tubes to ship.
Customers Who Bought This Also Bought