Shipper, EAS Frozen, Therapak®
Catalog #: 10029-420
Supplier: Therapak, LLC
Specifications
- Description:EAS frozen shipping system
- Cat. no.:10029-420
- Supplier No.:56885
Specifications
About this item
EAS shipping systems for frozen specimens feature Therapak®'s standard small insulated cooler (ID: 8 x 6 x 8”) and transport box (OD: 11 x 9 x 11”) with a Class 9 Dry Ice label, making it ideal for overnight shipments.
Includes specimen bag, absorbent material, Exempt Animal Specimen label, biohazard symbol label, and cushioning material. Add sample tubes and dry ice to ship.
Customers Who Bought This Also Bought