Shipper, EAS Refrigerated, Therapak®
Catalog #: 10029-418
Supplier: Therapak, LLC
Specifications
- Description:EAS refrigerated shipping systems
- Cat. no.:10029-418
- Supplier No.:56884
Specifications
About this item
EAS shipping systems for refrigerated specimens feature Therapak®'s standard small insulated cooler (ID: 8 x 6 x 8”) and transport box (OD: 11 x 9 x 11”), which is ideal for overnight shipments via common carriers such as DHL and FedEx
Refrigerated shipping system includes: 8-oz cold packs, specimen bag, absorbent material, Exempt Animal Specimen label, biohazard label and cushioning material. Each case includes one set of packing instructions.
Customers Who Bought This Also Bought