Shipper, EAS Ambient for 6 Tubes, Therapak®
Catalog #: 10029-414
Supplier: Therapak, LLC
Specifications
- Description:Medium transport box, specimen bag, up to 6 tubes
- Cat. no.:10029-414
- Supplier No.:56882
Specifications
About this item
Ambient shipping system features an Aqui-Pak segmented absorbent pouch to hold up to six standard blood or urine tubes.
Shipped with pre-assembled boxes, this transport shipper includes: medium transport box (ID: 7 x 4 x 4"; OD: 8 x 4.25 x 4.25"), specimen transport bag, absorbent material, Exempt Animal Specimen label, biohazard symbol label and cushioning material where applicable.
Customers Who Bought This Also Bought